missing translation for 'onlineSavingsMsg'
Learn More

5-Heptyn-3-ol 97.0+%, TCI America™

Catalog No. H04595ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04595ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04595ML Supplier TCI America Supplier No. H04595ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An acetylenic alcohol used as a chemical building block for synthetic organic research.

Chemical Identifiers

CAS 19781-82-9
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00046680
InChI Key VOHBMHAELZLACG-UHFFFAOYSA-N
PubChem CID 543129
IUPAC Name hept-5-yn-3-ol
SMILES CCC(CC#CC)O

Specifications

Color Yellow
Boiling Point 43°C
Quantity 5 mL
Formula Weight 112.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Heptyn-3-ol