accessibility menu, dialog, popup

UserName

CH3H3CHO

CAS RN 19781-82-9

IUPAC Name: hept-5-yn-3-ol
Synonyms: hept-5-yn-3-ol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: VOHBMHAELZLACG-UHFFFAOYNA-N
SMILES: CCC(O)CC#CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results