missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ 5 ft. Cell Logic™ Class II, Type A2 Biosafety Cabinet, US Model
Cell Logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensure the highest standard of safety and performance.
Supplier: Labconco™ 342591102
Description
Performance Features:
- Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection.
- Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load.
- Optimized airflow minimizes energy use and noise output to industry-leading levels.
- Logic OS with 5 in. line-of-sight touchscreen display.
- Access cabinet information such as filter life, alerts and alarms, data logs.
- Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
- Full, six-year warranty on all Class II biosafety cabinets.
Optional Configurations:
- Scope-Ready™ Package consisting of 7.0 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar.
- Stand-Still™ Isolation Platform is required (sold separately).
- Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. wide x 12.0 in. deep (27 x 30 cm) located on the left side of the work surface.
- Water circulator is required (not included).
- Telescoping base stand with shelf and leveling feet.
Chemical Identifiers
| 5 ft. Cell Logic Class II, Type A2 Biosafety Cabinet | |
| Stainless Steel (304) | |
| Class II, Type A2 Bio Safety Cabinet | |
| 63 dBA | |
| 240 W | |
| 61.9 in. | |
| 66.4 in. | |
| 5 in. touchscreen | |
| HEPA | |
| 50 W | |
| 157.2 cm | |
| 1 | |
| 6 Years | |
| 25.7 in. | |
| 27 in. | |
| 57.2 cm | |
| 32 in. | |
| 240 W |
| ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
| 153.7 cm | |
| Cell Logic | |
| 2 Duplex | |
| 2 Double, Left and Right Side | |
| 22.9 cm | |
| 892 m³/hr. | |
| 99.99% | |
| 168.7 cm | |
| LED, UV | |
| 9 in. | |
| 525 CFM | |
| 60.5 in. | |
| 22.5 in. | |
| 65.3 cm | |
| 69 cm | |
| 81.3 cm |
Specifications
| 5 ft. Cell Logic Class II, Type A2 Biosafety Cabinet | |
| Stainless Steel (304) | |
| Class II, Type A2 Bio Safety Cabinet | |
| 63 dBA | |
| North America (115 V, 20 A) | |
| 2 Double, Left and Right Side | |
| 22.9 cm | |
| 892 m³/hr. | |
| 99.99% | |
| 168.7 cm | |
| 50 W | |
| 157.2 cm | |
| 1 | |
| 6 Years | |
| 60 Hz | |
| 22.5 in. | |
| 65.3 cm | |
| 69 cm | |
| 81.3 cm | |
| 240 W |
| ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
| 153.7 cm | |
| Cell Logic | |
| 2 Duplex | |
| 240 W | |
| 61.9 in. | |
| 66.4 in. | |
| 5 in. touchscreen | |
| HEPA | |
| Cabinet, Accessory Package, Scope-Ready, Temp-Zone, Base Stand | |
| LED, UV | |
| 9 in. | |
| 525 CFM | |
| 60.5 in. | |
| 25.7 in. | |
| 27 in. | |
| 57.2 cm | |
| 32 in. | |
| 115 V |