missing translation for 'onlineSavingsMsg'
Learn More

5-Formylthiophene-2-carbonitrile 98.0+%, TCI America™

Catalog No. F11331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11331G 1 g
F11335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11331G Supplier TCI America Supplier No. F11331G

Chemical Identifiers

CAS 21512-16-3
Molecular Formula C6H3NOS
Molecular Weight (g/mol) 137.156
MDL Number MFCD09037804
InChI Key PZIFYWVUYHMYOA-UHFFFAOYSA-N
Synonym 5-cyano-2-thiophene carbaldehyde, 5-cyano-2-thiophenecarboxaldehyde, 5-cyanothiophene-2-carboxaldehyde, 2-thiophenecarbonitrile, 5-formyl, 5-cyano-2-thiophenecarbaldehyde, 2-formyl-5-cyanothiophen, 2-cyano-5-formylthiophene, 2-formyl-5-cyanothiophene, 2-cyano-5-formyl-thiophene, 5-cyanothiophene-2-aldehyde
PubChem CID 12280004
IUPAC Name 5-formylthiophene-2-carbonitrile
SMILES C1=C(SC(=C1)C#N)C=O

Specifications

Melting Point 93°C
Quantity 1 g
UN Number 3439
Formula Weight 137.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Formylthiophene-2-carbonitrile