accessibility menu, dialog, popup

UserName

ONOSO

CAS RN 21512-16-3

IUPAC Name: 5-nitrothiophene-2-carbaldehyde
Synonyms: 5-nitrothiophene-2-carbaldehyde
Molecular Weight (g/mol): 157.14
Molecular Formula: C5H3NO3S
InChi Key: CHTSWZNXEKOLPM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(S1)C=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)