missing translation for 'onlineSavingsMsg'
Learn More

5-Formylthiophene-2-boronic acid, 97%

Catalog No. AAH2928903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2928903 1 g
AAH2928906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2928903 Supplier Thermo Scientific Chemicals Supplier No. H2928903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4347-33-5 | C5H5BO3S | 155.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4347-33-5
Molecular Formula C5H5BO3S
Molecular Weight (g/mol) 155.96
MDL Number MFCD02093666
InChI Key DEQOVKFWRPOPQP-UHFFFAOYSA-N
Synonym 5-formyl-2-thiopheneboronic acid, 5-formylthiophen-2-boronic acid, 5-formylthiophen-2-yl boronic acid, 5-formylthiophene-2-boronic acid, 2-formylthiophene-5-boronic acid, 5-formylthienyl-2-boronic acid, 5-formyl-2-thienyl boronic acid, boronic acid, 5-formyl-2-thienyl, 5-formyl-2-thiopheneboronicacid, pubchem1722
PubChem CID 2773430
IUPAC Name (5-formylthiophen-2-yl)boronic acid
SMILES OB(O)C1=CC=C(S1)C=O

Specifications

Melting Point ∽124°C (decomposition)
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 155.97
Percent Purity 97%
Chemical Name or Material 5-Formylthiophene-2-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only