accessibility menu, dialog, popup

UserName

HOBOHSO

CAS RN 4347-33-5

IUPAC Name: (5-formylthiophen-2-yl)boronic acid
Synonyms: 5-formylthiophen-2-ylboronic acid
Molecular Weight (g/mol): 155.96
Molecular Formula: C5H5BO3S
InChi Key: DEQOVKFWRPOPQP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(S1)C=O
Quantity 
  • (3)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)