missing translation for 'onlineSavingsMsg'
Learn More

5-Formylthiazole 98.0+%, TCI America™

Catalog No. f09085g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09085G 5 g
F09081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09085G Supplier TCI America Supplier No. F09085G
Only null left

Chemical Identifiers

CAS 1003-32-3
Molecular Formula C4H3NOS
Molecular Weight (g/mol) 113.13
MDL Number MFCD02179516
InChI Key ZXRLWHGLEJGMNO-UHFFFAOYSA-N
Synonym thiazole-5-carboxaldehyde, thiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde, thiazole-5-carboxyaldehyde, 5-formylthiazole, 5-formyl-1,3-thiazole, 1,3-thiazole-5-carboxaldehyde, thiazole-5-aldehyde, zlchem 512, pubchem14219
PubChem CID 2773577
IUPAC Name 1,3-thiazole-5-carbaldehyde
SMILES O=CC1=CN=CS1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 113.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Formylthiazole