CAS RN 1003-32-3
IUPAC Name:
1,3-thiazole-5-carbaldehyde
Synonyms:
1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol):
113.13
Molecular Formula:
C4H3NOS
InChi Key:
ZXRLWHGLEJGMNO-UHFFFAOYSA-N
SMILES:
O=CC1=CN=CS1
1
–
1
of
1
results
1
5-Formylthiazole 98.0+%, TCI America™
CAS: 1003-32-3 Molecular Formula: C4H3NOS Molecular Weight (g/mol): 113.13 MDL Number: MFCD02179516 InChI Key: ZXRLWHGLEJGMNO-UHFFFAOYSA-N Synonym: thiazole-5-carboxaldehyde,thiazole-5-carbaldehyde,5-thiazolecarboxaldehyde,thiazole-5-carboxyaldehyde,5-formylthiazole,5-formyl-1,3-thiazole,1,3-thiazole-5-carboxaldehyde,thiazole-5-aldehyde,zlchem 512,pubchem14219 PubChem CID: 2773577 IUPAC Name: 1,3-thiazole-5-carbaldehyde SMILES: O=CC1=CN=CS1