missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2-thiophenecarboxylic Acid 98.0+%, TCI America™

$158.44 - $400.78

Chemical Identifiers

CAS 4565-31-5
Molecular Formula C6H3O3S
Molecular Weight (g/mol) 155.15
MDL Number MFCD00191437
InChI Key VFEAMMGYJFFXKV-UHFFFAOYSA-M
Synonym 5-Formylthenoic Acid
PubChem CID 818882
IUPAC Name 5-formylthiophene-2-carboxylate
SMILES [O-]C(=O)C1=CC=C(S1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F03361G
View Documents
TCI America
F03361G
1 g
Each for $158.44
Only null left
 
F03365G
View Documents
TCI America
F03365G
5 g
Each for $400.78
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4565-31-5
155.15
VFEAMMGYJFFXKV-UHFFFAOYSA-M
818882
[O-]C(=O)C1=CC=C(S1)C=O
C6H3O3S
MFCD00191437
5-Formylthenoic Acid
5-formylthiophene-2-carboxylate

Specifications

4565-31-5
C6H3O3S
1 g
VFEAMMGYJFFXKV-UHFFFAOYSA-M
5-formylthiophene-2-carboxylate
818882
≥98.0% (GC,T)
5-Formyl-2-thiophenecarboxylic Acid
169°C
MFCD00191437
5-Formylthenoic Acid
[O-]C(=O)C1=CC=C(S1)C=O
155.15
156.16
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents