missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Formyl-2-thiophenecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America F03365G
Specifications
| 5-Formyl-2-thiophenecarboxylic Acid | |
| 169°C | |
| MFCD00191437 | |
| 5-Formylthenoic Acid | |
| [O-]C(=O)C1=CC=C(S1)C=O | |
| 155.15 | |
| 156.16 | |
| Crystalline Powder |
| 4565-31-5 | |
| C6H3O3S | |
| 5 g | |
| VFEAMMGYJFFXKV-UHFFFAOYSA-M | |
| 5-formylthiophene-2-carboxylate | |
| 818882 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 4565-31-5 | |
| 155.15 | |
| VFEAMMGYJFFXKV-UHFFFAOYSA-M | |
| 818882 | |
| [O-]C(=O)C1=CC=C(S1)C=O |
| C6H3O3S | |
| MFCD00191437 | |
| 5-Formylthenoic Acid | |
| 5-formylthiophene-2-carboxylate |
Safety and Handling
TSCA : No