missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Formyl-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Chemical Name or Material | 5-Formyl-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride) |
| CAS | 127972-02-5 |
| Color | Yellow |
| Molecular Formula | C8H9BO4 |
| Quantity | 1 g |
| MDL Number | MFCD01319044 |
| Synonym | 2-methoxy-5-formylphenylboronic acid, 5-formyl-2-methoxyphenyl boronic acid, 5-formyl-2-methoxybenzeneboronic acid, 5-formyl-2-methoxy-phenyl boronic acid, 2-methoxy-5-formylbenzeneboronic acid, 3-borono-4-methoxybenzaldehyde, 3-dihydroxyboranyl-4-methoxybenzaldehyde, boronic acid, 5-formyl-2-methoxyphenyl, pubchem7991, acmc-209bd6 |
| InChI Key | NKKNXLPHCRLBDY-UHFFFAOYSA-N |
| SMILES | COC1=CC=C(C=O)C=C1B(O)O |
| IUPAC Name | (5-formyl-2-methoxyphenyl)boronic acid |
| Show More |