missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2,4-dimethyl-3-pyrrolecarboxylic Acid 98.0+%, TCI America™

Catalog No. F09395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09395G 5 g
F09391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09395G Supplier TCI America Supplier No. F09395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 253870-02-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD06202342
InChI Key YCIHQDVIAISDPS-UHFFFAOYSA-N
Synonym 5-formyl-2,4-dimethylpyrrole-3-carboxylic acid, 2,4-dimethyl-5-formylpyrrole-3-carboxylic acid, 3,5-dimethyl-2-formylpyrrole-4-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl, 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylicacid, 3,5-dimethyl-2-formyl-1h-pyrrole-4-carboxylic acid, 3,5-dimethyl-2-formylindole-4-carboxylic acid, zlchem 714, pubchem9414, acmc-209zxq
PubChem CID 11073792
IUPAC Name 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
SMILES CC1=C(NC(=C1C(=O)O)C)C=O

Specifications

Melting Point 283°C
Color Yellow
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylic Acid