missing translation for 'onlineSavingsMsg'
Learn More

5-Formamido-1-[2-(formyloxy)ethyl]pyrazole 97.0+%, TCI America™

Catalog No. F08905G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08905G Supplier TCI America Supplier No. F08905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116856-18-9
Molecular Formula C7H9N3O3
Molecular Weight (g/mol) 183.167
MDL Number MFCD09744040
InChI Key ADNBFDFIEWMUBJ-UHFFFAOYSA-N
Synonym 2-5-formamido-1h-pyrazol-1-yl ethyl formate, 5-formamide-1-2-formyloxyethl pyrazole, 5-formamido-1-2-formyloxyethyl pyrazole, 2-5-formamidopyrazol-1-yl ethyl formate, 5-formylamino-1-2-formyloxyethyl pyrazole, 5-formamido-1-2-formyloxy ethyl pyrazole, pubchem15369, tpc-i141, 5-formamido-l-2-formyloxyethyl pyrazole
PubChem CID 14996567
IUPAC Name 2-(5-formamidopyrazol-1-yl)ethyl formate
SMILES C1=C(N(N=C1)CCOC=O)NC=O

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 5 g
Formula Weight 183.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Formamido-1-[2-(formyloxy)ethyl]pyrazole