accessibility menu, dialog, popup

UserName

CAS RN 116856-18-9

IUPAC Name: 2-(5-formamido-1H-pyrazol-1-yl)ethyl formate
Synonyms: 2-(5-formamidopyrazol-1-yl)ethyl formate
Molecular Weight (g/mol): 183.17
Molecular Formula: C7H9N3O3
InChi Key: ADNBFDFIEWMUBJ-UHFFFAOYSA-N
SMILES: O=CNC1=CC=NN1CCOC=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results