missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil 99.0+%, TCI America™

Supplier:  TCI America F015125G

 View more versions of this product

Catalog No. F015125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Fluorouracil
283°C
C4H3FN2O2
25 g
5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton
FC1=CNC(=O)NC1=O
130.08
CHEBI:46345
≥99.0% (HPLC,T)
51-21-8
White
MFCD00006018
2811
GHASVSINZRGABV-UHFFFAOYSA-N
5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
3385
130.08
Crystalline Powder

Chemical Identifiers

51-21-8
130.08
GHASVSINZRGABV-UHFFFAOYSA-N
3385
5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
C4H3FN2O2
MFCD00006018
5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton
CHEBI:46345
FC1=CNC(=O)NC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1180

RTECSNumber : YR0350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions