missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil, 99%

Catalog No. p-7021215
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1345606 5 g
AAA1345614 25 g
AAA1345622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1345606 Supplier Thermo Scientific Chemicals Supplier No. A1345606

CAS: 51-21-8 | C4H3FN2O2 | 130.08 g/mol

5-Fluorouracil is used as an antitumor agent in the treatment of anal, breast, colorectal, oesophageal, stomach, pancreatic and skin cancers. It finds application as a suicide inhibitor due to its irreversible inhibition of thymidylate synthase. It is also used in the treatment of actinic keratoses and bowen's disease. Further, it serves as a potent antineoplastic agent in clinical use. In addition to this, it acts as a DNA synthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethylformamide, dimethyl sulfoxide, methanol, hot water, ammonium hydroxide, sodium hydroxide, ethanol and water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 51-21-8
Molecular Formula C4H3FN2O2
Molecular Weight (g/mol) 130.08
MDL Number MFCD00006018
InChI Key GHASVSINZRGABV-UHFFFAOYSA-N
Synonym 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton
PubChem CID 3385
ChEBI CHEBI:46345
IUPAC Name 5-fluoro-1H-pyrimidine-2,4-dione
SMILES FC1=CNC(=O)NC1=O

Specifications

Melting Point ∽282°C (decomposition)
Assay Percent Range 99%
Quantity 5 g
UN Number UN2811
Beilstein 127172
Sensitivity Light sensitive
Merck Index 14,4181
Solubility Information Soluble in dimethylformamide,dimethyl sulfoxide,methanol,hot water,ammonium hydroxide,sodium hydroxide,ethanol and water.
Formula Weight 130.08
Percent Purity 99%
Chemical Name or Material 5-Fluorouracil
Show More Show Less
Hazard Category H301-H312+H332-H315-H319-H335-H341-H361d
Hazard Statement GHS H Statement
H360-H341-H373-H302
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-085-6
RTECSNumber YR0350000
TSCA Yes
storageNote1 Air Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only