missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluorouracil, ∽99%, MP Biomedicals™
$449.32 - $449.32
Chemical Identifiers
| CAS | 51-21-8 |
|---|---|
| Molecular Formula | C4H3FN2O2 |
| Molecular Weight (g/mol) | 130.08 |
| MDL Number | MFCD00006018 |
| InChI Key | GHASVSINZRGABV-UHFFFAOYSA-N |
| Synonym | 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton |
| PubChem CID | 3385 |
| ChEBI | CHEBI:46345 |
| IUPAC Name | 5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | FC1=CNC(=O)NC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10172210
|
MP Biomedicals Inc
0210172210 |
10 g |
Each for $449.32
|
|
|||||
Chemical Identifiers
| 51-21-8 | |
| 130.08 | |
| GHASVSINZRGABV-UHFFFAOYSA-N | |
| 3385 | |
| 5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| C4H3FN2O2 | |
| MFCD00006018 | |
| 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton | |
| CHEBI:46345 | |
| FC1=CNC(=O)NC1=O |
Specifications
| 51-21-8 | |
| ∽99% | |
| MFCD00006018 | |
| 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton | |
| FC1=CNC(=O)NC1=O | |
| 130.08 | |
| CHEBI:46345 | |
| Powder |
| White | |
| C4H3FN2O2 | |
| 10 g | |
| GHASVSINZRGABV-UHFFFAOYSA-N | |
| 5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 3385 | |
| 130.08 | |
| 5-Fluorouracil |