missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Fluorouracil, 99%

Catalog No. AC228440050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228440050 5 g Glass bottle
AC228440010 1 g Glass bottle
AC228440250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC228440050 Supplier Thermo Scientific Chemicals Supplier No. 228440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-Fluorouracil, CAS # 51-21-8, is an analogue base composed of uracil with a position 5 hydrogen fluorine substitution. | CAS: 51-21-8 | C4H3FN2O2 | 130.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 5-Fluorouracil is an antimetabolite and antineoplastic that prevents DNA synthesis by blocking conversion of deoxyuridylic acid

• 5-Fluorouracil can replace uracil in RNA and prevent protein synthesis and cell growth

Applications

• 5-Fluorouracil can be used for laboratory research on tumor growth and immunosuppression

• 5-Fluorouracil may also be appropriate for laboratory studies of neurotoxicity, cell lysis, and DNA damage

Chemical Identifiers

CAS 51-21-8
Molecular Formula C4H3FN2O2
Molecular Weight (g/mol) 130.08
MDL Number MFCD00006018
InChI Key GHASVSINZRGABV-UHFFFAOYSA-N
Synonym 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton
PubChem CID 3385
ChEBI CHEBI:46345
IUPAC Name 5-fluoro-1H-pyrimidine-2,4-dione
SMILES FC1=CNC(=O)NC1=O

Specifications

Melting Point 278.0°C to 282.0°C
Color White
Boiling Point 190.0°C to 200.0°C (0.1 mmHg)
Loss on Drying 0.5% (80°C, 4 hrs) (in vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 24,IV,1229
Merck Index 15,4213
Solubility Information Solubility in water: 12.2g/L (20°C). Other solubilities: 2.9g/L (20°C) in ethanol,readily soluble in aqueous alkali solutions,soluble in dmso
Formula Weight 130.08
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorouracil
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
Suspected of damaging the unborn child.
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irri
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comforta
EINECSNumber 200-085-6

RUO – Research Use Only