missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluorosalicylic Acid 98.0+%, TCI America™
$87.60 - $253.51
Chemical Identifiers
| CAS | 345-16-4 |
|---|---|
| Molecular Formula | C7H5FO3 |
| Molecular Weight (g/mol) | 156.112 |
| MDL Number | MFCD00002456 |
| InChI Key | JWPRICQKUNODPZ-UHFFFAOYSA-N |
| Synonym | 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol |
| PubChem CID | 67658 |
| IUPAC Name | 5-fluoro-2-hydroxybenzoic acid |
| SMILES | C1=CC(=C(C=C1F)C(=O)O)O |
Chemical Identifiers
| 345-16-4 | |
| 156.112 | |
| JWPRICQKUNODPZ-UHFFFAOYSA-N | |
| 67658 | |
| C1=CC(=C(C=C1F)C(=O)O)O |
| C7H5FO3 | |
| MFCD00002456 | |
| 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol | |
| 5-fluoro-2-hydroxybenzoic acid |
Specifications
| 345-16-4 | |
| C7H5FO3 | |
| 1 g | |
| JWPRICQKUNODPZ-UHFFFAOYSA-N | |
| 5-fluoro-2-hydroxybenzoic acid | |
| 67658 | |
| ≥98.0% (GC,T) | |
| 5-Fluorosalicylic Acid |
| 180°C | |
| MFCD00002456 | |
| 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol | |
| C1=CC(=C(C=C1F)C(=O)O)O | |
| 156.112 | |
| 156.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No