missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorosalicylic Acid 98.0+%, TCI America™

Catalog No. F04691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04691G 1 g
F04695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04691G Supplier TCI America Supplier No. F04691G

Chemical Identifiers

CAS 345-16-4
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.112
MDL Number MFCD00002456
InChI Key JWPRICQKUNODPZ-UHFFFAOYSA-N
Synonym 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol
PubChem CID 67658
IUPAC Name 5-fluoro-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1F)C(=O)O)O

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 156.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorosalicylic Acid