Learn More
5-Fluorosalicylic acid, 97%
CAS: 345-16-4 | C7H5FO3 | 156.112 g/mol
$130.27 - $391.90
Chemical Identifiers
| CAS | 345-16-4 |
|---|---|
| Molecular Formula | C7H5FO3 |
| Molecular Weight (g/mol) | 156.112 |
| MDL Number | MFCD00002456 |
| InChI Key | JWPRICQKUNODPZ-UHFFFAOYSA-N |
| Synonym | 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol |
| PubChem CID | 67658 |
| IUPAC Name | 5-fluoro-2-hydroxybenzoic acid |
| SMILES | C1=CC(=C(C=C1F)C(=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2175103
|
Thermo Scientific Chemicals
B2175103 |
1 g |
Each for $130.27
|
|
|||||
|
AAB2175106
|
Thermo Scientific Chemicals
B2175106 |
5 g |
Each for $391.90
|
|
|||||
Description
5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 345-16-4 | |
| 156.112 | |
| JWPRICQKUNODPZ-UHFFFAOYSA-N | |
| 67658 | |
| C1=CC(=C(C=C1F)C(=O)O)O |
| C7H5FO3 | |
| MFCD00002456 | |
| 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol | |
| 5-fluoro-2-hydroxybenzoic acid |
Specifications
| 345-16-4 | |
| C7H5FO3 | |
| 1 g | |
| 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol | |
| JWPRICQKUNODPZ-UHFFFAOYSA-N | |
| 5-fluoro-2-hydroxybenzoic acid | |
| 67658 | |
| 97% |
| 178°C to 182°C | |
| MFCD00002456 | |
| 2209120 | |
| Slightly soluble in water. | |
| C1=CC(=C(C=C1F)C(=O)O)O | |
| 156.112 | |
| 156.11 | |
| 5-Fluorosalicylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-455-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only