missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluororesorcinol 98.0+%, TCI America™
$616.05 - $1853.93
Chemical Identifiers
| CAS | 75996-29-1 |
|---|---|
| Molecular Formula | C6H5FO2 |
| Molecular Weight (g/mol) | 128.102 |
| MDL Number | MFCD08457182 |
| InChI Key | LYFBZGKZAZBANN-UHFFFAOYSA-N |
| PubChem CID | 12658778 |
| IUPAC Name | 5-fluorobenzene-1,3-diol |
| SMILES | C1=C(C=C(C=C1O)F)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
F1081200MG
|
TCI America
F1081200MG |
200 mg |
Each for $616.05
|
|
|||||
|
F10811G
|
TCI America
F10811G |
1 g |
Each for $1,853.93
|
|
|||||
Chemical Identifiers
| 75996-29-1 | |
| 128.102 | |
| LYFBZGKZAZBANN-UHFFFAOYSA-N | |
| 5-fluorobenzene-1,3-diol |
| C6H5FO2 | |
| MFCD08457182 | |
| 12658778 | |
| C1=C(C=C(C=C1O)F)O |
Specifications
| 75996-29-1 | |
| C6H5FO2 | |
| 200 mg | |
| C1=C(C=C(C=C1O)F)O | |
| 128.102 | |
| 128.10 | |
| Crystalline Powder |
| 138°C | |
| MFCD08457182 | |
| LYFBZGKZAZBANN-UHFFFAOYSA-N | |
| 5-fluorobenzene-1,3-diol | |
| 12658778 | |
| ≥98.0% (GC,T) | |
| 5-Fluororesorcinol |
Safety and Handling
TSCA : No