Learn More
5-Fluoropyridine-3-boronic acid, 98%
CAS: 872041-86-6 | C5H5BFNO2 | 140.908 g/mol
Supplier: Thermo Scientific Chemicals H5286003
Description
5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.
Solubility
Soluble in hot methanol.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
| 5-Fluoropyridine-3-boronic acid | |
| 233°C to 236°C | |
| MFCD07368243 | |
| 5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine | |
| FVEDGBRHTGXPOK-UHFFFAOYSA-N | |
| (5-fluoropyridin-3-yl)boronic acid | |
| 44717403 | |
| 98% |
| 872041-86-6 | |
| C5H5BFNO2 | |
| 1 g | |
| Soluble in hot methanol. | |
| B(C1=CC(=CN=C1)F)(O)O | |
| 140.908 | |
| 140.91 |
Chemical Identifiers
| 872041-86-6 | |
| 140.908 | |
| FVEDGBRHTGXPOK-UHFFFAOYSA-N | |
| 44717403 | |
| B(C1=CC(=CN=C1)F)(O)O |
| C5H5BFNO2 | |
| MFCD07368243 | |
| 5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine | |
| (5-fluoropyridin-3-yl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only