missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoropyridine-3-boronic acid, 98%

Catalog No. p-7035146
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52860MD 250 mg
AAH5286003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52860MD Supplier Thermo Scientific Chemicals Supplier No. H52860MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 872041-86-6 | C5H5BFNO2 | 140.908 g/mol

5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 872041-86-6
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.908
MDL Number MFCD07368243
InChI Key FVEDGBRHTGXPOK-UHFFFAOYSA-N
Synonym 5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine
PubChem CID 44717403
IUPAC Name (5-fluoropyridin-3-yl)boronic acid
SMILES B(C1=CC(=CN=C1)F)(O)O

Specifications

Melting Point 233°C to 236°C
Quantity 250 mg
Solubility Information Soluble in hot methanol.
Formula Weight 140.91
Percent Purity 98%
Chemical Name or Material 5-Fluoropyridine-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only