missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorooxindole 98.0+%, TCI America™

Catalog No. F06991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06991G 1 g
F06995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06991G Supplier TCI America Supplier No. F06991G

Chemical Identifiers

CAS 56341-41-4
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD02179598
InChI Key DDIIYGHHUMKDGI-UHFFFAOYSA-N
Synonym 5-fluorooxindole, 5-fluoroindolin-2-one, 5-fluoro-2-oxindole, 5-fluoro-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one, 5-fluoro-1,3-dihydro, 5-fluoro-1,3-dihydro-indol-2-one, 5-fluoro-2-indolinone, 5-fluoro-2,3-dihydro-1h-indol-2-one, 5-fluoro indoline-2-one, 5-fluoro-2,3-dihydro-2-oxoindole
PubChem CID 3731012
IUPAC Name 5-fluoro-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)F)NC1=O

Specifications

Melting Point 145°C
Quantity 1 g
Formula Weight 151.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorooxindole