accessibility menu, dialog, popup

UserName

FHNO

CAS RN 56341-41-4

IUPAC Name: 5-fluoro-2,3-dihydro-1H-indol-2-one
Synonyms: 5-fluoro-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: DDIIYGHHUMKDGI-UHFFFAOYSA-N
SMILES: FC1=CC=C2NC(=O)CC2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results