missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindoline, 97%

Catalog No. p-7035928
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Catalog No. Quantity
AAH54158MD 250 mg
AAH5415803 1 g
AAH5415806 5 g
3 options

Catalog No. AAH54158MD

Supplier: Thermo Scientific Chemicals H54158MD

Only null left

CAS: 2343-22-8 | C8H8FN | 137.157 g/mol

5-Fluoroindoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoroindoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 2343-22-8
Molecular Formula C8H8FN
Molecular Weight (g/mol) 137.157
MDL Number MFCD00214461
InChI Key NXQRMQIYCWFDGP-UHFFFAOYSA-N
PubChem CID 2774463
IUPAC Name 5-fluoro-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)F

Specifications

Quantity 250 mg
Sensitivity Air sensitive
Synonym 5-fluoroindoline, 5-fluoro-2,3-dihydro-1h-indole, 1h-indole,5-fluoro-2,3-dihydro, 1h-indole, 5-fluoro-2,3-dihydro, 5-fluoro-indoline, peakdale1_000398, 5-fluoro-2,3-dihydro-1h-indol, 2,3-dihydro-5-fluoro-1h-indole
Solubility Information Slightly soluble in water.
Formula Weight 137.16
Percent Purity 97%
Chemical Name or Material 5-Fluoroindoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only