CAS RN 2343-22-8
IUPAC Name:
5-fluoro-2,3-dihydro-1H-indole
Synonyms:
5-fluoro-2,3-dihydro-1H-indole
Molecular Weight (g/mol):
137.16
Molecular Formula:
C8H8FN
InChi Key:
NXQRMQIYCWFDGP-UHFFFAOYSA-N
SMILES:
FC1=CC2=C(NCC2)C=C1
1
–
1
of
1
results
1
Promotions Available
5-Fluoroindoline, 97%
5-Fluoroindoline, 97%
CAS: 2343-22-8 Molecular Formula: C8H8FN Molecular Weight (g/mol): 137.157 MDL Number: MFCD00214461 InChI Key: NXQRMQIYCWFDGP-UHFFFAOYSA-N PubChem CID: 2774463 IUPAC Name: 5-fluoro-2,3-dihydro-1H-indole SMILES: C1CNC2=C1C=C(C=C2)F