missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129850
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07161G 1 g
F07165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07161G Supplier TCI America Supplier No. F07161G
Only null left

Chemical Identifiers

CAS 399-76-8
Molecular Formula C9H6FNO2
Molecular Weight (g/mol) 179.15
MDL Number MFCD00005612
InChI Key WTXBRZCVLDTWLP-UHFFFAOYSA-N
Synonym 5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043
PubChem CID 1820
IUPAC Name 5-fluoro-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1F)C=C(N2)C(=O)O

Specifications

Melting Point 249°C
Color White-Yellow
Quantity 1 g
Formula Weight 179.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoroindole-2-carboxylic Acid