Learn More
5-Fluoroindole-2-carboxylic acid, 98+%
CAS: 399-76-8 | C9H6FNO2 | 179.15 g/mol
Supplier: Thermo Scientific Chemicals A1279406
Description
5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Specifications
| 5-Fluoroindole-2-carboxylic acid | |
| ∼259°C (decomposition) | |
| MFCD00005612 | |
| 153217 | |
| It is highly soluble in water. | |
| C1=CC2=C(C=C1F)C=C(N2)C(=O)O | |
| 179.15 | |
| 179.15 |
| 399-76-8 | |
| C9H6FNO2 | |
| 5 g | |
| 5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043 | |
| WTXBRZCVLDTWLP-UHFFFAOYSA-N | |
| 5-fluoro-1H-indole-2-carboxylic acid | |
| 1820 | |
| ≥98% |
Chemical Identifiers
| 399-76-8 | |
| 179.15 | |
| WTXBRZCVLDTWLP-UHFFFAOYSA-N | |
| 1820 | |
| C1=CC2=C(C=C1F)C=C(N2)C(=O)O |
| C9H6FNO2 | |
| MFCD00005612 | |
| 5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043 | |
| 5-fluoro-1H-indole-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-919-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only