missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole-2-carboxylic acid, 98+%

Catalog No. AAA1279403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1279403 1 g
AAA1279406 5 g
AAA1279414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1279403 Supplier Thermo Scientific Chemicals Supplier No. A1279403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 399-76-8 | C9H6FNO2 | 179.15 g/mol

5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is highly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.

Chemical Identifiers

CAS 399-76-8
Molecular Formula C9H6FNO2
Molecular Weight (g/mol) 179.15
MDL Number MFCD00005612
InChI Key WTXBRZCVLDTWLP-UHFFFAOYSA-N
Synonym 5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043
PubChem CID 1820
IUPAC Name 5-fluoro-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1F)C=C(N2)C(=O)O

Specifications

Melting Point ∼259°C (decomposition)
Quantity 1 g
Beilstein 153217
Solubility Information It is highly soluble in water.
Formula Weight 179.15
Percent Purity ≥98%
Chemical Name or Material 5-Fluoroindole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-919-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only