missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-o-tolyl Isothiocyanate 98.0+%, TCI America™

Catalog No. F05155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05155G 5 g
F051525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05155G Supplier TCI America Supplier No. F05155G
Only null left

Chemical Identifiers

CAS 175205-39-7
Molecular Formula C8H6FNS
Molecular Weight (g/mol) 167.201
InChI Key QVCYTKXGZLOVFA-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylphenyl isothiocyanate, 2-methyl-5-fluorophenyl isothiocyanate, 5-fluoro-o-tolyl isothiocyanate, 5-fluoro-2-methylphenylisothiocyanate, isothiocyanic acid 5-fluoro-o-tolyl ester, acmc-20am3d, 4-fluoro-2-isothiocyanatotoluene, 2-methyl-5-fluorophenylisothiocyanate, 5-fluoro-2-methylbenzenisothiocyanate, 5-fluoro-2-methylphenyl-isothiocyanate
PubChem CID 519443
IUPAC Name 4-fluoro-2-isothiocyanato-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)N=C=S

Specifications

Color Yellow
Quantity 5 g
Formula Weight 167.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Fluoro-o-tolyl Isothiocyanate