missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-o-anisaldehyde 97.0+%, TCI America™

Catalog No. p-7129680
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
F04281G 1 g
F04285G 5 g
2 options

Catalog No. F04281G

Supplier: TCI America F04281G

Only null left

Chemical Identifiers

CAS 19415-51-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143458
InChI Key CRLDWFVRQNUUSZ-UHFFFAOYSA-N
PubChem CID 2734943
IUPAC Name 5-fluoro-2-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)F)C=O

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 118°C
Quantity 1 g
Synonym 2-methoxy-5-fluorobenzaldehyde, 5-fluoro-o-anisaldehyde, 5-fluoro-o-methoxybenzaldehyde, benzaldehyde, 5-fluoro-2-methoxy, 5-fluoro-2-methoxy-benzaldehyde, 5-fluoro-o-2-methoxybenzaldehyde, pubchem2636, acmc-209exz, 2-methoxy-5-fluoro benzaldehyde, benzaldehyde,5-fluoro-2-methoxy
Formula Weight 154.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Fluoro-o-anisaldehyde