missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-m-xylene, 97%
CAS: 461-97-2 | C8H9F | 124.158 g/mol
$264.79 - $264.79
Chemical Identifiers
| CAS | 461-97-2 |
|---|---|
| Molecular Formula | C8H9F |
| Molecular Weight (g/mol) | 124.158 |
| MDL Number | MFCD00052366 |
| InChI Key | RCWIWNUVHNAUQC-UHFFFAOYSA-N |
| Synonym | 3,5-dimethylfluorobenzene, 5-fluoro-m-xylene, 1,3-dimethyl-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethyl, pubchem4410, 5-fluoro-1,3-xylene, acmc-1ahss, 3,5-dimethyl-fluorobenzene, 3,5-dimethylphenyl fluoride, ksc497s5r |
| PubChem CID | 521192 |
| IUPAC Name | 1-fluoro-3,5-dimethylbenzene |
| SMILES | CC1=CC(=CC(=C1)F)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1502006
|
Thermo Scientific Chemicals
A1502006 |
5 g |
Each for $264.79
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 461-97-2 | |
| 124.158 | |
| RCWIWNUVHNAUQC-UHFFFAOYSA-N | |
| 521192 | |
| CC1=CC(=CC(=C1)F)C |
| C8H9F | |
| MFCD00052366 | |
| 3,5-dimethylfluorobenzene, 5-fluoro-m-xylene, 1,3-dimethyl-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethyl, pubchem4410, 5-fluoro-1,3-xylene, acmc-1ahss, 3,5-dimethyl-fluorobenzene, 3,5-dimethylphenyl fluoride, ksc497s5r | |
| 1-fluoro-3,5-dimethylbenzene |
Specifications
| 461-97-2 | |
| 144°C to 145°C | |
| 1.476 | |
| MFCD00052366 | |
| 1926374 | |
| RCWIWNUVHNAUQC-UHFFFAOYSA-N | |
| 1-fluoro-3,5-dimethylbenzene | |
| 521192 | |
| 97% |
| 0.983 | |
| 5 g | |
| C8H9F | |
| UN1993 | |
| 3,5-dimethylfluorobenzene, 5-fluoro-m-xylene, 1,3-dimethyl-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethyl, pubchem4410, 5-fluoro-1,3-xylene, acmc-1ahss, 3,5-dimethyl-fluorobenzene, 3,5-dimethylphenyl fluoride, ksc497s5r | |
| CC1=CC(=CC(=C1)F)C | |
| 124.158 | |
| 124.16 | |
| 5-Fluoro-m-xylene |
Safety and Handling
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only