missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-m-xylene, 97%

Catalog No. AAA1502006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1502006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1502006 Supplier Thermo Scientific Chemicals Supplier No. A1502006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 461-97-2 | C8H9F | 124.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 461-97-2
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.158
MDL Number MFCD00052366
InChI Key RCWIWNUVHNAUQC-UHFFFAOYSA-N
Synonym 3,5-dimethylfluorobenzene, 5-fluoro-m-xylene, 1,3-dimethyl-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethyl, pubchem4410, 5-fluoro-1,3-xylene, acmc-1ahss, 3,5-dimethyl-fluorobenzene, 3,5-dimethylphenyl fluoride, ksc497s5r
PubChem CID 521192
IUPAC Name 1-fluoro-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)F)C

Specifications

Density 0.983
Quantity 5 g
Refractive Index 1.476
UN Number UN1993
Beilstein 1926374
Boiling Point 144°C to 145°C
Formula Weight 124.16
Percent Purity 97%
Chemical Name or Material 5-Fluoro-m-xylene
Hazard Category H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only