missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals

$236.55 - $236.55

Chemical Identifiers

CAS 243459-91-8
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.102
MDL Number MFCD00236302
InChI Key QWLZSSYHAJSEHU-UHFFFAOYSA-N
PubChem CID 2774803
IUPAC Name 5-fluoro-2-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1F)O)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3244506
View Documents
Thermo Scientific Chemicals
H3244506
5 g
Each for $236.55
Only null left
 
Description

Description

As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.

Solubility
Sparingly Soluble (0.40 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

243459-91-8
180.102
QWLZSSYHAJSEHU-UHFFFAOYSA-N
5-fluoro-2-(trifluoromethyl)phenol
C7H4F4O
MFCD00236302
2774803
C1=CC(=C(C=C1F)O)C(F)(F)F

Specifications

243459-91-8
C7H4F4O
5 g
5-fluoro-2-trifluoromethyl phenol, 5-fluoro-2-trifluoromethyl-phenol, 4-fluoro-2-hydroxybenzotrifluoride, pubchem2171, intermediates-zcf02657, phenol,5-fluoro-2-trifluoromethyl, phenol, 5-fluoro-2-trifluoromethyl
QWLZSSYHAJSEHU-UHFFFAOYSA-N
5-fluoro-2-(trifluoromethyl)phenol
2774803
97%
48°C to 52°C
MFCD00236302
UN3261
Sparingly Soluble (0.40g/L) (25°C).
C1=CC(=C(C=C1F)O)C(F)(F)F
180.102
180.1
5-Fluoro-2-(trifluoromethyl)phenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only