missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-2-nitrotoluene 98.0+%, TCI America™
Supplier: TCI America F030125G
Specifications
| 5-Fluoro-2-nitrotoluene | |
| White-Yellow | |
| C7H6FNO2 | |
| 25 g | |
| JHFOWEGCZWLHNW-UHFFFAOYSA-N | |
| 4-fluoro-2-methyl-1-nitrobenzene | |
| 94956 | |
| ≥98.0% (GC) |
| 446-33-3 | |
| 98°C | |
| MFCD00007283 | |
| 5-fluoro-2-nitrotoluene, 3-fluoro-6-nitrotoluene, 4-fluoro-2-methylnitrobenzene, 2-nitro-5-fluoro toluene, benzene, 4-fluoro-2-methyl-1-nitro, toluene, 5-fluoro-2-nitro, 2-nitro-5-fluorotoluene, 2-methyl-4-fluoronitrobenzene, 3-methyl-4-nitrofluorobenzene, 4-fluoro-2-methyl-1-nitro-benzene | |
| CC1=C(C=CC(=C1)F)[N+](=O)[O-] | |
| 155.128 | |
| 155.13 | |
| Crystals |
Chemical Identifiers
| 446-33-3 | |
| 155.128 | |
| JHFOWEGCZWLHNW-UHFFFAOYSA-N | |
| 94956 | |
| CC1=C(C=CC(=C1)F)[N+](=O)[O-] |
| C7H6FNO2 | |
| MFCD00007283 | |
| 5-fluoro-2-nitrotoluene, 3-fluoro-6-nitrotoluene, 4-fluoro-2-methylnitrobenzene, 2-nitro-5-fluoro toluene, benzene, 4-fluoro-2-methyl-1-nitro, toluene, 5-fluoro-2-nitro, 2-nitro-5-fluorotoluene, 2-methyl-4-fluoronitrobenzene, 3-methyl-4-nitrofluorobenzene, 4-fluoro-2-methyl-1-nitro-benzene | |
| 4-fluoro-2-methyl-1-nitrobenzene |
Safety and Handling
TSCA : No