missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrotoluene, 98+%

Catalog No. AAA1394409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1394409 10 g
AAA1394418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1394409 Supplier Thermo Scientific Chemicals Supplier No. A1394409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 446-33-3 | C7H6FNO2 | 155.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 446-33-3
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007283
InChI Key JHFOWEGCZWLHNW-UHFFFAOYSA-N
Synonym 5-fluoro-2-nitrotoluene, 3-fluoro-6-nitrotoluene, 4-fluoro-2-methylnitrobenzene, 2-nitro-5-fluoro toluene, benzene, 4-fluoro-2-methyl-1-nitro, toluene, 5-fluoro-2-nitro, 2-nitro-5-fluorotoluene, 2-methyl-4-fluoronitrobenzene, 3-methyl-4-nitrofluorobenzene, 4-fluoro-2-methyl-1-nitro-benzene
PubChem CID 94956
IUPAC Name 4-fluoro-2-methyl-1-nitrobenzene
SMILES CC1=C(C=CC(=C1)F)[N+](=O)[O-]

Specifications

Melting Point ∼20°C
Density 1.27
Boiling Point 97°C to 98°C (10 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.528
Quantity 10 g
UN Number UN2810
Beilstein 2046652
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2-nitrotoluene
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 207-164-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only