missing translation for 'onlineSavingsMsg'
Learn More

(5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate 95.0+%, TCI America™

Catalog No. F1110200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1110200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1110200MG Supplier TCI America Supplier No. F1110200MG
Only null left

Chemical Identifiers

CAS 1868173-33-4
Molecular Formula C22H21FINO8S
Molecular Weight (g/mol) 605.372
InChI Key CADOMGBBYLEKSI-UHFFFAOYSA-M
Synonym (5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium Tosylate
PubChem CID 131697916
IUPAC Name (5-fluoro-2-nitrophenyl)-(2,4,6-trimethoxyphenyl)iodanium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].COC1=CC(=C(C(=C1)OC)[I+]C2=C(C=CC(=C2)F)[N+](=O)[O-])OC

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 605.37
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator