missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrophenol, 99%

Catalog No. AC241970250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241970250 25 g Glass bottle
AC241971000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC241970250 Supplier Thermo Scientific Chemicals Supplier No. 241970250

CAS: 446-36-6 | C6H4FNO3 | 157.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 446-36-6
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD00007107
InChI Key QQURWFRNETXFTN-UHFFFAOYSA-N
Synonym 2-nitro-5-fluorophenol, 3-fluoro-6-nitrophenol, phenol, 5-fluoro-2-nitro, 4-fluoro-2-hydroxynitrobenzene, 5-fluoro-2-nitro-phenol, pubchem1490, pubchem2371, acmc-209jzc, 3-fluoro 6-nitrophenol, 5-flouro-2-nitrophenol
PubChem CID 9937
IUPAC Name 5-fluoro-2-nitrophenol
SMILES C1=CC(=C(C=C1F)O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 32°C to 35°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(NO2)OH
Quantity 25 g
Beilstein 06, II, 225
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 157.1
Percent Purity 99%
Physical Form Powder, Crystals or Crystalline Mass and/or Chunks
Chemical Name or Material 5-Fluoro-2-nitrophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-168-6

RUO – Research Use Only