missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrobenzoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 398870050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC398870050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

320-98-9
185.11
GHYZIXDKAPMFCS-UHFFFAOYSA-N
2737420
C7H4FNO4
MFCD00055635
5-fluoro-2-nitrobenzoic acid, 2-nitro-5-fluorobenzoic acid, 3-fluoro-6-nitrobenzoic acid, 5-fluoro-2-nitro-benzoic acid, benzoic acid, 5-fluoro-2-nitro, 2-carboxy-4-fluoronitrobenzene, pubchem1309, acmc-1aiv3, 5-fluoro-2nitrobenzoic acid, 5-fluor-2-nitrobenzoic acid
C1=CC(=C(C=C1F)C(=O)O)[N+](=O)[O-]

Specifications

5-Fluoro-2-nitrobenzoic acid
97.5
Authentic
Glass bottle
5g
5-fluoro-2-nitrobenzoic acid, 2-nitro-5-fluorobenzoic acid, 3-fluoro-6-nitrobenzoic acid, 5-fluoro-2-nitro-benzoic acid, benzoic acid, 5-fluoro-2-nitro, 2-carboxy-4-fluoronitrobenzene, pubchem1309, acmc-1aiv3, 5-fluoro-2nitrobenzoic acid, 5-fluor-2-nitrobenzoic acid
C1=CC(=C(C=C1F)C(=O)O)[N+](=O)[O-]
2737420
98%
320-98-9
100.0
97.5% min. (HPLC)
C7H4FNO4
MFCD00055635
GHYZIXDKAPMFCS-UHFFFAOYSA-N
185.11
185.11
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents