missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-2-nitrobenzaldehyde 98.0+%, TCI America™
$85.75 - $315.03
Chemical Identifiers
| CAS | 395-81-3 |
|---|---|
| Molecular Formula | C7H4FNO3 |
| Molecular Weight (g/mol) | 169.11 |
| MDL Number | MFCD00153175 |
| InChI Key | KKAFVHUJZPVWND-UHFFFAOYSA-N |
| Synonym | 2-nitro-5-fluorobenzaldehyde, 5-fluoro-2-nitrobenzadehyde, benzaldehyde, 5-fluoro-2-nitro, 4-fluoro-2-formylnitrobenzene, 5-fluoro-2-nitro-benzaldehyde, 5-fluioro-2-nitrobenzaldehyde, pubchem3078, acmc-209j6a, 3-fluoro-6-nitrobenzaldehyde, ksc581k6t |
| PubChem CID | 587090 |
| IUPAC Name | 5-fluoro-2-nitrobenzaldehyde |
| SMILES | [O-][N+](=O)C1=CC=C(F)C=C1C=O |
Chemical Identifiers
| 395-81-3 | |
| 169.11 | |
| KKAFVHUJZPVWND-UHFFFAOYSA-N | |
| 587090 | |
| [O-][N+](=O)C1=CC=C(F)C=C1C=O |
| C7H4FNO3 | |
| MFCD00153175 | |
| 2-nitro-5-fluorobenzaldehyde, 5-fluoro-2-nitrobenzadehyde, benzaldehyde, 5-fluoro-2-nitro, 4-fluoro-2-formylnitrobenzene, 5-fluoro-2-nitro-benzaldehyde, 5-fluioro-2-nitrobenzaldehyde, pubchem3078, acmc-209j6a, 3-fluoro-6-nitrobenzaldehyde, ksc581k6t | |
| 5-fluoro-2-nitrobenzaldehyde |
Specifications
| 395-81-3 | |
| 153°C | |
| MFCD00153175 | |
| 2-nitro-5-fluorobenzaldehyde, 5-fluoro-2-nitrobenzadehyde, benzaldehyde, 5-fluoro-2-nitro, 4-fluoro-2-formylnitrobenzene, 5-fluoro-2-nitro-benzaldehyde, 5-fluioro-2-nitrobenzaldehyde, pubchem3078, acmc-209j6a, 3-fluoro-6-nitrobenzaldehyde, ksc581k6t | |
| [O-][N+](=O)C1=CC=C(F)C=C1C=O | |
| 169.11 | |
| 169.11 | |
| Crystalline Powder |
| 94°C | |
| C7H4FNO3 | |
| 5 g | |
| KKAFVHUJZPVWND-UHFFFAOYSA-N | |
| 5-fluoro-2-nitrobenzaldehyde | |
| 587090 | |
| ≥98.0% (GC) | |
| 5-Fluoro-2-nitrobenzaldehyde |
Safety and Handling
TSCA : No