missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrobenzaldehyde 98.0+%, TCI America™

Catalog No. F06455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06455G 5 g
F064525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06455G Supplier TCI America Supplier No. F06455G
Only null left

Chemical Identifiers

CAS 395-81-3
Molecular Formula C7H4FNO3
Molecular Weight (g/mol) 169.11
MDL Number MFCD00153175
InChI Key KKAFVHUJZPVWND-UHFFFAOYSA-N
Synonym 2-nitro-5-fluorobenzaldehyde, 5-fluoro-2-nitrobenzadehyde, benzaldehyde, 5-fluoro-2-nitro, 4-fluoro-2-formylnitrobenzene, 5-fluoro-2-nitro-benzaldehyde, 5-fluioro-2-nitrobenzaldehyde, pubchem3078, acmc-209j6a, 3-fluoro-6-nitrobenzaldehyde, ksc581k6t
PubChem CID 587090
IUPAC Name 5-fluoro-2-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC=C(F)C=C1C=O

Specifications

Melting Point 94°C
Boiling Point 153°C
Quantity 5 g
Formula Weight 169.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-nitrobenzaldehyde