missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitroanisole 98.0+%, TCI America™

Catalog No. p-7129842
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07035G 5 g
F070325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07035G Supplier TCI America Supplier No. F07035G

Chemical Identifiers

CAS 448-19-1
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.127
MDL Number MFCD00077541
InChI Key WLKUSVNHZXUEFO-UHFFFAOYSA-N
Synonym 4-Fluoro-2-methoxy-1-nitrobenzene
PubChem CID 2779250
IUPAC Name 4-fluoro-2-methoxy-1-nitrobenzene
SMILES COC1=C(C=CC(=C1)F)[N+](=O)[O-]

Specifications

Melting Point 49°C
Color Yellow
Boiling Point 272°C
Quantity 5 g
Formula Weight 171.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-nitroanisole