missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylindole 98.0+%, TCI America™

Catalog No. p-7130047
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10471G 1 g
F10475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10471G Supplier TCI America Supplier No. F10471G

Chemical Identifiers

CAS 399-72-4
Molecular Formula C9H8FN
Molecular Weight (g/mol) 149.168
MDL Number MFCD02093649
InChI Key JJIUISYYTFDATN-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylindole, 1h-indole, 5-fluoro-2-methyl, zlchem 457, pubchem2158, 2-methyl-5-fluoroindole, 5-fluoro-2-methyl indole, 5-fluoro-2-methyl-indole, ksc222g8n
PubChem CID 2778715
IUPAC Name 5-fluoro-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)F

Specifications

Melting Point 100°C
Color Yellow
Boiling Point 120°C
Quantity 1 g
Formula Weight 149.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-methylindole