accessibility menu, dialog, popup

UserName

H3CHNF

CAS RN 399-72-4

IUPAC Name: 5-fluoro-2-methyl-1H-indole
Synonyms: 5-fluoro-2-methyl-1H-indole
Molecular Weight (g/mol): 149.17
Molecular Formula: C9H8FN
InChi Key: JJIUISYYTFDATN-UHFFFAOYSA-N
SMILES: CC1=CC2=CC(F)=CC=C2N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)