missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylindole, 95%, Thermo Scientific Chemicals

Catalog No. p-7056908
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6612103 1 g
AAH6612106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6612103 Supplier Thermo Scientific Chemicals Supplier No. H6612103
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 399-72-4
Molecular Formula C9H8FN
Molecular Weight (g/mol) 149.168
MDL Number MFCD02093649
InChI Key JJIUISYYTFDATN-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylindole, 1h-indole, 5-fluoro-2-methyl, zlchem 457, pubchem2158, 2-methyl-5-fluoroindole, 5-fluoro-2-methyl indole, 5-fluoro-2-methyl-indole, ksc222g8n
PubChem CID 2778715
IUPAC Name 5-fluoro-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)F

Specifications

Melting Point 98°C to 101°C
Quantity 1 g
Beilstein 116627
Formula Weight 149.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Fluoro-2-methylindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 609-764-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only