missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzoxazole 97.0+%, TCI America™

Catalog No. F04205G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04205G 5 g
F04201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04205G Supplier TCI America Supplier No. F04205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 701-16-6
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD02093471
InChI Key GYRZYFSJULFMCS-UHFFFAOYSA-N
PubChem CID 2769350
IUPAC Name 5-fluoro-2-methyl-1,3-benzoxazole
SMILES CC1=NC2=C(O1)C=CC(=C2)F

Specifications

Boiling Point 90°C
Quantity 5 g
Formula Weight 151.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Fluoro-2-methylbenzoxazole