accessibility menu, dialog, popup

UserName

CAS RN 701-16-6

IUPAC Name: 5-fluoro-2-methyl-1,3-benzoxazole
Synonyms: 5-fluoro-2-methyl-1,3-benzoxazole
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: GYRZYFSJULFMCS-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(F)=CC=C2O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results